Order Entry
United States
ContactUsLinkComponent
Human Recombinant PSA/KLK3/APS (from HEK cells)
Human Recombinant PSA/KLK3/APS (from HEK cells)
Supplier:  ReVacc Scientific
New Product
Certificates
undefined
Human Recombinant PSA/KLK3/APS (from HEK cells)
Supplier:  ReVacc Scientific
New Product

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK cells
  • Species:
    Human
  • Tag sequence:
    Myc tag and His tag
  • Storage Conditions:
    −80 °C
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Prostate-Specific Antigen|Semenogelase|Seminin|Gamma-Seminoprotein|P-30 Antigen|Kallikrein-3
  • Protein/Peptide Name:
    PSA/KLK3/APS
  • Molecular Weight:
    ~33.2 kDa
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Frozen liquid. In PBS pH 7.5
  • Nuclease-free:
    N
  • Tested Applications:
    SDS-PAGE, ELISA

Specifications

Frequently Bought Together