Anti-NiV Human Recombinant Monoclonal Antibody [clone: RV41]
Supplier: ReVacc Scientific
New Product
Certificates
About this item
This antibody has been tested to bind to Nipah Virus Glycoprotein (Catalog # 77761-744, 77761-746) with great affinity.
Type: Primary
Antigen: NiV
Clonality: Recombinant Monoclonal
Clone: RV41
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 77761-694
- 77761-692
- Antigen Symbol:
- NiV
- NiV
- Antigen Name:
- Nipah Virus
- Nipah Virus
- Antigen Synonyms:
- Glycoprotein (G)
- Glycoprotein (G)
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV41
- RV41
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Molecular Weight:
- 150 kDa
- 150 kDa
- Purification:
- Purified by Protein A chromatography
- Purified by Protein A chromatography
- Concentration:
- 1 mg/ml by Refractometry
- 1 mg/ml by Refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice
Specifications
Frequently Bought Together