Anti-His tag Bovine Recombinant Monoclonal Antibody [clone: RV7]
Supplier: ReVacc Scientific
New Product
Certificates
About this item
This antibody has been tested to bind to His tagged protein.
Type: Primary
Antigen: His tag
Clonality: Recombinant Monoclonal
Clone: RV7
Conjugation: Unconjugated
Epitope: 6-His (HHHHHH) or 8-His or 10-His tag
Host: Bovine
Isotype: IgG1 kappa
Reactivity: Bovine
Specifications
- Catalog No:
- 77761-644
- 77761-646
- Antigen Symbol:
- His tag
- His tag
- Antigen Name:
- Polyhistidine-tag
- Polyhistidine-tag
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV7
- RV7
- Reactivity:
- Bovine
- Bovine
- Host:
- Bovine
- Bovine
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Epitope:
- 6-His (HHHHHH) or 8-His or 10-His tag
- 6-His (HHHHHH) or 8-His or 10-His tag
- Purification:
- Purified by Protein C chromatography
- Purified by Protein C chromatography
- Concentration:
- 1 mg/ml by Refractometry
- 1 mg/ml by Refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice
Specifications
Frequently Bought Together