ContactUsLinkComponent
Hanna 1382 mg/L (ppm) TDS Calibration Solution, Hanna
Catalog # 470357-560
Supplier:  HANNA INSTRUMENTS
New Product
CAS Number:  
Hanna 1382 mg/L (ppm) TDS Calibration Solution, Hanna
Catalog # 470357-560
Supplier:  HANNA INSTRUMENTS
Supplier Number:  HI70032P
New Product
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    1382 mg/L (ppm) TDS calibration solution
  • Size:
    20 ml
  • Contents:
    25 tests
Specifications
About this item
NIST traceable and contained in portable single-use sachets.
  • Temperature chart of the actual µS/cm value at various temperatures printed on each bottle
  • Convenient packaging for one time use
Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened.
Expiration date of 5 years from the date of manufacture for an unopened sachet.