ContactUsLinkComponent
Hanna 1413 µS/cm Conductivity Standard
Hanna 1413 µS/cm Conductivity Standard
Catalog # 470357-558
Supplier:  HANNA INSTRUMENTS
New Product
CAS Number:  
Hanna 1413 µS/cm Conductivity Standard
Catalog # 470357-558
Supplier:  HANNA INSTRUMENTS
Supplier Number:  HI70031P
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    1413 µS/cm conductivity standard
  • Size:
    20 ml
  • Environmentally Preferable:
  • Contents:
    25 tests
Specifications
About this item
NIST traceable and contained in portable single-use sachets.
  • Temperature chart of the actual µS/cm value at various temperatures printed on each bottle
  • Convenient packaging for one time use
Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened.
Ordering information:
Expiration date of five years from the date of manufacture for an unopened sachet.