Nanalysis-100 Benchtop NMR Spectrometers, Nanalysis
New Product
The Nanalysis-100 benchtop NMR Spectrometers are market-leading, compact 100 MHz, 2.35 T, all-in-one spectrometers that are both easy-to-use and easy-to-site.
Market leading performance
- Multi-nuclear
- 1D and 2D experiments
- All-in-one enclosure
- Open API
- Remote access
-
Optional gradient based experiments
Accessible, affordable, automatable
At 100 MHz, this family of NMR spectrometers offers the highest resolution at the market with an easy to use interface. Acquire high-performance multi-nuclear 1D and/or 2D data at the touch of a button.
:
CE mark, CSA mark and FCC Declaration of Conformity.
:
Protect your benchtop NMR and peace of mind with up to four years of extended warranty.
Frequently Bought Together