Orders LinkContactUsLinkComponent
Proteometer-L Reactor
Proteometer-L Reactor
Proteometer-L reactor
Catalog # 77775-518
Supplier:   NOVILYTIC, LLC
Supplier #:  RCT-PL-001
Proteometer-L Reactor
Catalog # 77775-518
Supplier:  NOVILYTIC, LLC
Proteometer-L Reactor
Proteometer-L Reactor
Proteometer-L reactor
Price
$2,508.80
Log in for availability
Each
Product Details & Documents
Proteometer-L Reactor sold individually.
  • Proteometer-L Reactor replaces the LC column in the analytical system
Intended for use with the Proteometer-L kit.
Ordering information: Reorder individually as needed.
Packaging: Supplied in a custom box with instructions for use. Follow instructions in "Instructions for Use" for proper storage.
Specifications
  • Description:
    Proteometer-L reactor