VWR® 100 Conductivity Pocket Tester (Conductivity/TDS/Res./Sal.), Avantor
Catalog # 77618-946
Supplier: VWR International
New Product
Specifications
- Model:E-EC100
- Parameter:Conductivity/TDS/Salinity/Resistivity
Specifications
About this item
The VWR 100-series conductivity pocket tester is a complete system for conductivity measurements on the go.
High resolution LCD display
- Auto and manual HOLD function: Freezes readings on the display for easy and accurate recording
- IP67 waterproof
- Automatic temperature compensation capabilities
- Backlight adjustment and automatic shutdown: Extends battery service life
- Replaceable electrode
The easy to use VWR-EC100 Pocket Tester provides the flexibility of measuring conductivity, TDS (total dissolved solids), resistivity and salinity all with a single tester. The rugged carrying case allows you to easily transport the pocket tester, buffers and accessories to any location. Easily switch out the electrode to help prolong the life of the pocket tester.
Delivery:
Tester includes: Carrying case, wrist strap, conductivity standards (1413 µS/cm and 10.85 mS/cm), calibration bottles, AAA batteries (4).