Proteometer-L Reagent
77775-514
New Product
- Description:Proteometer-L Reagent
- Includes:Crimped and sealed reagent vials for Proteometer-L kit
Reagent vials for Proteometer-L kit.
Specifically formulated reagent vials for use with the Proteometer-L kit
Replacement reagent vials allow users to reorder when reactor and/or buffers are not required
Intended for use with the Proteometer-L kit
Crimped, sealed, sold in sets of 7.
:
Store at −20 °C.