Order Entry
United States
ContactUsLinkComponent
Anti-Biotin Monoclonal Antibody [clone: 1D4-C5] (TotalSeq™-A0268)
Anti-Biotin Monoclonal Antibody [clone: 1D4-C5] (TotalSeq™-A0268)
Catalog # 78027-465
Supplier:  Biolegend
New Product
Anti-Biotin Monoclonal Antibody [clone: 1D4-C5] (TotalSeq™-A0268)
Catalog # 78027-465
Supplier:  Biolegend
Supplier Number:  409019
New Product

Specifications

  • Antibody Type:
    Secondary
  • Antigen Symbol:
    Biotin
  • Clonality:
    Monoclonal
  • Clone:
    1D4-C5
  • Conjugation:
    TotalSeq™-A0268
  • Isotype:
    IgG2a kappa
  • Size:
    10 μg
  • Storage Temperature:
    2 -8 °C
  • Concentration:
    0.50 mg/ml
  • Shipping Temperature:
    Ambient RT

Specifications

About this item

TotalSeq™-A0268 anti-Biotin [1D4-C5]; Isotype: Mouse IgG2a, κ; Reactivity: ; Apps: PG

Each lot of this is quality control tested by https://www.biolegend.com/protocols/cell-surface-flow-cytometry-staining-protocol/4283/immunofluorescent staining with flow cytometric analysis and the oligomer sequence is confirmed by sequencing. TotalSeq™-A antibodies are compatible with 10x Genomics Single Cell Gene Expression https://www.10xgenomics.com/solutions/single-cell/#product-listSolutions.; To maximize performance, it is strongly recommended that the reagent be titrated for each application, and that you centrifuge the dilution before adding to the cells at 14,000xg at 2 - 8°C for 10 minutes. Carefully pipette out the liquid avoiding the bottom of the tube and add to the cell suspension. For Proteogenomics analysis, the suggested starting amount of this reagent for titration is ≤ 1.0 µg per million cells in 100 µL volume. Refer to the corresponding TotalSeq™ protocol for specific staining instructions.;

Antibody type: Secondary
Antigen symbol: Biotin
Clonality: Monoclonal
Clone: 1D4-C5
Conjugation: TotalSeq™-A0268
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse