- Antibody Type:Control
- Antigen Symbol:IgG2a kappa
- Clonality:Monoclonal
- Clone:MOPC-173
- Conjugation:TotalSeq™-B1472
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Size:10 μg
- Storage Temperature:2 -8 °C
- Concentration:0.50 mg/ml
- Shipping Temperature:Ambient RT
TotalSeq™-B1472 Mouse IgG2a, κ isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Reactivity: ; Apps: ICPG
Each lot of this is quality control tested by https://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/ intracellular immunofluorescent staining with flow cytometric analysis and the oligomer sequence is confirmed by sequencing. TotalSeq™-B antibodies are compatible with 10x Genomics Single Cell Gene Expression https://www.10xgenomics.com/solutions/single-cell/#product-listSolutions.; To maximize performance, it is strongly recommended that the reagent be titrated for each application, and that you centrifuge the dilution before adding to the cells at 14,000xg at 2 - 8°C for 10 minutes. Carefully pipette out the liquid avoiding the bottom of the tube and add to the cell suspension. For Proteogenomics analysis, the suggested starting amount of this reagent for titration is ≤ 0.5 µg per million cells in 100 µL volume. Refer to the corresponding TotalSeq™ protocol for specific staining instructions.;
Antibody type: Control
Antigen symbol: IgG2a kappa
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: TotalSeq™-B1472
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: