- Antibody Type:Primary
- Antigen Symbol:CXCL10
- Clonality:Monoclonal
- Clone:J034D6
- Conjugation:TotalSeq™-C1207
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Size:10 μg
- Gene ID:3627
- Storage Temperature:2 -8 °C
- Concentration:0.50 mg/ml
- Shipping Temperature:Ambient RT
TotalSeq™-C1207 anti-human CXCL10 (IP-10) [J034D6]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICPG
Each lot of this is quality control tested by https://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/intracellular immunofluorescent staining with flow cytometric analysis and the oligomer sequence is confirmed by sequencing. TotalSeq™-C antibodies are compatible with https://www.10xgenomics.com/10x Genomics Single Cell Gene Expression Solutions.; To maximize performance, it is strongly recommended that the reagent be titrated for each application, and that you centrifuge the dilution before adding to the cells at 14,000xg at 2 - 8°C for 10 minutes. Carefully pipette out the liquid avoiding the bottom of the tube and add to the cell suspension. For Proteogenomics analysis, the suggested starting amount of this reagent for titration is ≤ 0.5 µg per million cells in 100 µL volume. Refer to the corresponding TotalSeq™ protocol for specific staining instructions.;
Antibody type: Primary
Antigen symbol: CXCL10
Clonality: Monoclonal
Clone: J034D6
Conjugation: TotalSeq™-C1207
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Human