Order Entry
United States
ContactUsLinkComponent
Anti-IFN-gamma Monoclonal Antibody [clone: B27] (TotalSeq™-C1003)
Anti-IFN-gamma Monoclonal Antibody [clone: B27] (TotalSeq™-C1003)
Catalog # 78027-499
Supplier:  Biolegend
New Product
Anti-IFN-gamma Monoclonal Antibody [clone: B27] (TotalSeq™-C1003)
Catalog # 78027-499
Supplier:  Biolegend
Supplier Number:  506551
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    IFN-gamma
  • Clonality:
    Monoclonal
  • Clone:
    B27
  • Conjugation:
    TotalSeq™-C1003
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Size:
    10 μg
  • Gene ID:
    3458
  • Storage Temperature:
    2 -8 °C
  • Concentration:
    0.50 mg/ml
  • Shipping Temperature:
    Ambient RT

Specifications

About this item

TotalSeq™-C1003 anti-human IFN-γ [B27]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICPG

Each lot of this is quality control tested by https://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/intracellular immunofluorescent staining with flow cytometric analysis and the oligomer sequence is confirmed by sequencing. TotalSeq™-C antibodies are compatible with https://www.10xgenomics.com/10x Genomics Single Cell Gene Expression Solutions.; To maximize performance, it is strongly recommended that the reagent be titrated for each application, and that you centrifuge the dilution before adding to the cells at 14,000xg at 2 - 8°C for 10 minutes. Carefully pipette out the liquid avoiding the bottom of the tube and add to the cell suspension. For Proteogenomics analysis, the suggested starting amount of this reagent for titration is ≤ 0.5 µg per million cells in 100 µL volume. Refer to the corresponding TotalSeq™ protocol for specific staining instructions.;

Antibody type: Primary
Antigen symbol: IFN-gamma
Clonality: Monoclonal
Clone: B27
Conjugation: TotalSeq™-C1003
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human