Order Entry
United States
ContactUsLinkComponent
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2] (TotalSeq™-C1149)
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2] (TotalSeq™-C1149)
Catalog # 78027-493
Supplier:  Biolegend
New Product
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2] (TotalSeq™-C1149)
Catalog # 78027-493
Supplier:  Biolegend
Supplier Number:  500849
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    IL-4
  • Clonality:
    Monoclonal
  • Clone:
    MP4-25D2
  • Conjugation:
    TotalSeq™-C1149
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Size:
    10 μg

Specifications

About this item

TotalSeq™-C1149 anti-human IL-4 [MP4-25D2]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ICPG

Each lot of this is quality control tested by https://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/intracellular immunofluorescent staining with flow cytometric analysis and the oligomer sequence is confirmed by sequencing. TotalSeq™-C antibodies are compatible with https://www.10xgenomics.com/10x Genomics Single Cell Gene Expression Solutions.; To maximize performance, it is strongly recommended that the reagent be titrated for each application, and that you centrifuge the dilution before adding to the cells at 14,000xg at 2 - 8°C for 10 minutes. Carefully pipette out the liquid avoiding the bottom of the tube and add to the cell suspension. For Proteogenomics analysis, the suggested starting amount of this reagent for titration is ≤ 0.5 µg per million cells in 100 µL volume. Refer to the corresponding TotalSeq™ protocol for specific staining instructions.; Buyer is solely responsible for determining whether Buyer has all intellectual property rights that are necessary for Buyer's intended uses of the BioLegend TotalSeq™ products. For example, for any technology platform Buyer uses with TotalSeq™, it is Buyer's sole responsibility to determine whether it has all necessary third-party intellectual property rights to use that platform and TotalSeq™ with that platform.;

Antibody type: Primary
Antigen symbol: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: TotalSeq™-C1149
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human