Order Entry
United States
ContactUsLinkComponent
 
Anti-GM-CSF Rat Monoclonal Antibody [clone: BVD2-21C11] (TotalSeq™-C0943)
Catalog # 78027-497
Supplier:  Biolegend
New Product
Anti-GM-CSF Rat Monoclonal Antibody [clone: BVD2-21C11] (TotalSeq™-C0943)
Catalog # 78027-497
Supplier:  Biolegend
Supplier Number:  502325
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    GM-CSF
  • Clonality:
    Monoclonal
  • Clone:
    BVD2-21C11
  • Conjugation:
    TotalSeq™-C0943
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Size:
    10 μg
  • Gene ID:
    1437
  • Storage Temperature:
    2 -8 °C
  • Concentration:
    0.50 mg/ml
  • Shipping Temperature:
    Ambient RT

Specifications

About this item

TotalSeq™-C0943 anti-human GM-CSF [BVD2-21C11]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ICPG

Each lot of this is quality control tested by https://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/intracellular immunofluorescent staining with flow cytometric analysis and the oligomer sequence is confirmed by sequencing. TotalSeq™-C antibodies are compatible with https://www.10xgenomics.com/10x Genomics Single Cell Gene Expression Solutions.; To maximize performance, it is strongly recommended that the reagent be titrated for each application, and that you centrifuge the dilution before adding to the cells at 14,000xg at 2 - 8°C for 10 minutes. Carefully pipette out the liquid avoiding the bottom of the tube and add to the cell suspension. For Proteogenomics analysis, the suggested starting amount of this reagent for titration is ≤ 0.5 µg per million cells in 100 µL volume. Refer to the corresponding TotalSeq™ protocol for specific staining instructions. ;

Antibody type: Primary
Antigen symbol: GM-CSF
Clonality: Monoclonal
Clone: BVD2-21C11
Conjugation: TotalSeq™-C0943
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human