Order Entry
United States
ContactUsLinkComponent
Anti-IL-3 Rat Monoclonal Antibody [clone: BVD3-1F9] (TotalSeq™-B1161)
Anti-IL-3 Rat Monoclonal Antibody [clone: BVD3-1F9] (TotalSeq™-B1161)
Catalog # 78027-492
Supplier:  Biolegend
New Product
Anti-IL-3 Rat Monoclonal Antibody [clone: BVD3-1F9] (TotalSeq™-B1161)
Catalog # 78027-492
Supplier:  Biolegend
Supplier Number:  500609
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    IL-3
  • Clonality:
    Monoclonal
  • Clone:
    BVD3-1F9
  • Conjugation:
    TotalSeq™-B1161
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Size:
    10 μg

Specifications

About this item

TotalSeq™-B1161 anti-human IL-3 [BVD3-1F9]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ICPG

Each lot of this is quality control tested by ihttps://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/ntracellular immunofluorescent staining with flow cytometric analysis and the oligomer sequence is confirmed by sequencing. TotalSeq™- B antibodies are compatible with 10x Genomics Single Cell Gene Expression https://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/Solutions.; To maximize performance, it is strongly recommended that the reagent be titrated for each application, and that you centrifuge the dilution before adding to the cells at 14,000xg at 2 - 8°C for 10 minutes. Carefully pipette out the liquid avoiding the bottom of the tube and add to the cell suspension. For Proteogenomics analysis, the suggested starting amount of this reagent for titration is ≤ 0.5 µg per million cells in 100 µL volume. Refer to the corresponding TotalSeq™ protocol for specific staining instructions. ;

Antibody type: Primary
Antigen symbol: IL-3
Clonality: Monoclonal
Clone: BVD3-1F9
Conjugation: TotalSeq™-B1161
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human