Order Entry
United States
ContactUsLinkComponent
Anti-CXCL5 Monoclonal Antibody [clone: J111B7] (TotalSeq™-B1209)
Anti-CXCL5 Monoclonal Antibody [clone: J111B7] (TotalSeq™-B1209)
Catalog # 78027-510
Supplier:  Biolegend
New Product
Anti-CXCL5 Monoclonal Antibody [clone: J111B7] (TotalSeq™-B1209)
Catalog # 78027-510
Supplier:  Biolegend
Supplier Number:  524107
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CXCL5
  • Clonality:
    Monoclonal
  • Clone:
    J111B7
  • Conjugation:
    TotalSeq™-B1209
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Size:
    10 μg
  • Gene ID:
    6374
  • Storage Temperature:
    2 -8 °C
  • Concentration:
    0.50 mg/ml
  • Shipping Temperature:
    Ambient RT

Specifications

About this item

TotalSeq™-B1209 anti-human CXCL5 [J111B7]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICPG

Each lot of this is quality control tested by https://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/intracellular immunofluorescent staining with flow cytometric analysis and the oligomer sequence is confirmed by sequencing. TotalSeq™-B antibodies are compatible with 10x Genomics Single Cell Gene Expression https://www.biolegend.com/protocols/intracellular-flow-cytometry-staining-protocol/4260/Solutions.; To maximize performance, it is strongly recommended that the reagent be titrated for each application, and that you centrifuge the dilution before adding to the cells at 14,000xg at 2 - 8°C for 10 minutes. Carefully pipette out the liquid avoiding the bottom of the tube and add to the cell suspension. For Proteogenomics analysis, the suggested starting amount of this reagent for titration is ≤ 0.5 µg per million cells in 100 µL volume. Refer to the corresponding TotalSeq™ protocol for specific staining instructions.;

Antibody type: Primary
Antigen symbol: CXCL5
Clonality: Monoclonal
Clone: J111B7
Conjugation: TotalSeq™-B1209
Epitope:
Host:
Isotype: IgG2b kappa
Reactivity: Human