Order Entry
United States
ContactUsLinkComponent
Human IL-1 beta ELISPOT Kit (with un-coated plates)
Human IL-1 beta ELISPOT Kit (with un-coated plates)
Supplier:  Abcam
New Product
Certificates
Human IL-1 beta ELISPOT Kit (with un-coated plates)
Supplier:  Abcam
New Product

Specifications

  • Species Reactivity:
    Human
  • Sample Type:
    Suspension cells
  • Storage Temperature:
    4 °C
  • Processing Time:
    Multiple steps
  • Antigen Symbol:
    IL-1 beta
  • Regulatory Status:
    RUO

Specifications

Frequently Bought Together