Rat Interferon gamma ELISPOT Kit (with un-coated plates)
: Abcam
New Product
Rat Interferon gamma ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Rat Interferon gamma production in a single cell suspension.
- Species Reactivity:Rat
- Sample Type:Suspension cells
- Antigen Name:Interferon gamma
- Storage Temperature:4 °C
- Processing Time:Multiple steps
Frequently Bought Together