Pyruvate dehydrogenase (PDH) Protein Quantity Dipstick Assay Kit
Catalog # ABCA_AB109883-30TE
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Pyruvate dehydrogenase (PDH) Protein Quantity Dipstick Assay kit
- Size:30 tests
- Species Reactivity:Other;Human;Mouse;Rat
- Antigen Name:Pyruvate dehydrogenase
- Storage Temperature:4 °C
- Processing Time:Multiple steps
- Regulatory Status:RUO
- Format:Pre-coated
- Method:Sandwich
Specifications
About this item
Pyruvate dehydrogenase (PDH) Protein Quantity Dipstick Assay Kit is an immunoassay kit for the detection of Pyruvate dehydrogenase (PDH) Protein Quantity in Mouse, Rat, Cow, Human in Cell culture extracts, Tissue samples.