Human IL-2 ELISPOT Kits (with un-coated plates)
Supplier: Abcam
New Product
Certificates
About this item
Frequently Bought Together
VWR® Stripping Buffers
Supplier: VWR
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Western Blot Boxes
Supplier: Alkali Scientific
Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Blue Prestained Protein Standard, Broad Range (11 to 250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)