Human Granzyme B ELISPOT Kit (with un-coated plates)
Supplier: Abcam
New Product
Certificates
About this item
Human Granzyme B ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Human Granzyme B production in a single cell suspension.
Specifications
- Species Reactivity:Human
- Sample Type:Suspension cells
- Form:Uncoated
- Storage Temperature:+4 °C
- Processing Time:Multiple steps
- Antigen Symbol:Granzyme B
- Regulatory Status:RUO
Specifications
Frequently Bought Together