Order Entry
United States
ContactUsLinkComponent
 
Human IL-6 ELISPOT Kit (with un-coated plates)
 :  Abcam
New Product
 
undefined
Human IL-6 ELISPOT Kit (with un-coated plates)
 :  Abcam
New Product

 

  • Species Reactivity:
    Human
  • Sample Type:
    Suspension cells
  • Storage Temperature:
    4 °C
  • Processing Time:
    Multiple steps
  • Antigen Symbol:
    IL-6
  • Regulatory Status:
    RUO

 

Frequently Bought Together