Human TNF alpha ELISPOT Kit (with un-coated plates)
Supplier: Abcam
New Product
Certificates
About this item
Human TNF alpha ELISPOT kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Human TNF alpha production in a single cell suspension.
Specifications
- Species Reactivity:Human, Monkey
- Sample Type:Suspension cells
- Storage Temperature:4 °C
- Processing Time:Multiple steps
- Antigen Symbol:TNF alpha
- Regulatory Status:RUO
Specifications
Frequently Bought Together