Mouse Interferon gamma ELISPOT Kit (with un-coated plates)
Supplier: Abcam
New Product
Certificates
About this item
Mouse Interferon gamma ELISPOT kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Mouse Interferon gamma production in a single cell suspension.
Specifications
- Species Reactivity:Mouse
- Sample Type:Suspension cells
- Antigen Name:Interferon gamma
- Storage Temperature:+4 ℃
- Processing Time:Multiple steps
- Regulatory Status:RUO
Specifications
Frequently Bought Together