Human Interferon gamma + IL-10 ELISPOT Set
Catalog # ABCA_AB48722-20X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Human Interferon gamma + IL-10 ELISPOT set
- Size:20×96 tests
- Species Reactivity:Human
- Sample Type:Suspension cells
- Form:Uncoated
- Storage Temperature:+4 °C
- Processing Time:Multiple steps
- Antigen Symbol:Interferon gamma + IL10
- Regulatory Status:RUO
Specifications
About this item
Human Interferon gamma + IL-10 ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Interferon gamma + IL-10 production in a single cell suspension.