Order Entry
United States
ContactUsLinkComponent
Mouse IgG Isotyping Kit
Mouse IgG Isotyping Kit
Supplier:  Abcam
New Product
Certificates
undefined
Mouse IgG Isotyping Kit
Supplier:  Abcam
New Product

Specifications

  • Species Reactivity:
    Species independent
  • Storage Temperature:
    4 °C
  • Processing Time:
    Multiple steps
  • Regulatory Status:
    RUO

Specifications

Frequently Bought Together