Specifications
- Pack type:Glass bottle
- Description:AP-StabilPLUS
- Size:100 ml
Specifications
About this item
AP-StabilPLUS, ready-to-use (RTU)
- AP-StabilPLUS is a ready-to use buffer that appears as an opaque solution. The product is based on a mild Tris buffer containing proprietary stabilizing components. AP-StabilPLUS contains neither BSA, nor other material from bovine serum, azide, mercury or other toxic components.
AP-StabilPLUS is used for the dilution and stabilization of Alkaline Phosphatase (AP) labeled proteins and antibodies, in order to maintain the molecular conformation and prevent loss of activity over time. This enables the making of prediluted, ready-to-use AP-conjugates, minimizing assay errors in dilution.When testing the AP-StabilPLUS with AP conjugated antibodies, stability is seen as follows: at least 3 years at 2…8 °C at least 2 years at room temperature at least 4 weeks at 37 °C Stabilization of AP conjugated antibodies works well in low as well as in high protein dilutions.
Working Procedure 1.Make a series of dilutions of the AP-conjugated protein in AP-StabilPLUS in order to determine the optimal dilution. 2.Run the assay as usual or store the diluted APconjugated protein preferably at 2…8 °C. Tips and Tricks Avoid using phosphate buffers for AP-conjugated antibody assays. We recommend the use of Tris/HCl, Tween as the washing buffer, instead of a PBS buffer which will reduce signal significantly. Handling and Storage Store solution at 2…8 °C