About this item
Product Describtion:A dipeptide with an inhibitory effect on the excitability of the TAN. FY is one of the ACE-inhibitory dipeptides found in fermented milk. Administration of the dipeptide reduced blood pressure in spontaneously hypertensive rats.
- Density: ~1.3 g/cm³ Boiling Point: ~642.2 °C(1013 hPa) Melting Point: 320.45 °C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:17355-18-9
- Storage Conditions:Refrigerator
- Protein Synonyms:L-Phenylalanyl-L-tyrosine
- Protein/Peptide Name:H-Phe-Tyr-OH
- Purity:≥98%
- Molecular Weight:328.36 g/mol
- Molecular Formula:C₁₈H₂₀N₂O₄