ContactUsLinkComponent
RPMI Medium 1640 1X HEPES without L-Glutamine
New Product
undefined
RPMI Medium 1640 1X HEPES without L-Glutamine
New Product
  • Calcium:
    Yes
  • Magnesium:
    Yes
  • HEPES:
    Yes
  • Sodium bicarbonate:
    Yes
  • Sodium pyruvate:
    No
  • Phenol red:
    Yes
  • Stable glutamine:
    No
  • L-Glutamine:
    No
  • Glucose level:
    High
  • Media Base:
    RPMI 1640 - Roswell Park Memorial Institute Medium
Frequently Bought Together