Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Media Format:Liquid
- Pack type:Bottle
- Size:1 L
- Calcium:No
- Magnesium:No
- HEPES:No
- Sodium bicarbonate:No
- Sodium pyruvate:No
- Phenol red:No
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:Without
- Media Base:Proprietary Mixture (protein-free/chemically-defined)
- Application:Supports T cell expansion in static and dynamic,automation systems while maintaining functionality
Eliminates the adverse effects undefined components cause on T cell phenotypes
Supports polarization to targeted T cell types, such as Th1, Th2, cytotoxic T cells, to further possible therapy applications
Performs transduction of T cells without requiring spinoculation or transduction enhancers
Specifications
About this item
Chemically defined and animal component-free medium for T cell culture. PRIME-XV T Cell CDM supports viable T cell expansion across different culture vessels.
- Chemically defined, animal component-free formula Does not contain antibiotics or phenol red
PRIME-XV T Cell CDM is the first commercially-available, chemically defined, animal component-free medium for the expansion and transduction of human T cells. The formulation is optimized to deliver consistently vigorous growth while maintaining T cell functionality and potency. Designed and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management.