ContactUsLinkComponent
Ham's F12
Ham's F12
Catalog # 77975-315
New Product
CAS Number:  
undefined
Ham's F12
Catalog # 77975-315
Supplier Number:  9058-500ML
New Product
CAS Number:  
Specifications
  • Media Format:
    Liquid
  • Size:
    500 ml
  • Calcium:
    Yes
  • Magnesium:
    Yes
  • HEPES:
    No
  • Sodium bicarbonate:
    Yes
  • Sodium pyruvate:
    Yes
  • Phenol red:
    Yes
  • Stable glutamine:
    No
  • L-Glutamine:
    No
  • Glucose level:
    High
  • Media Base:
    Ham's F12 Nutrient Mixture
Specifications
About this item
Ham's F12 was originally developed for use in cloning Chinese hamster ovary and lung cells. This medium is useful with serum supplements for a variety of cell culture applications, including the growth of myeloma and hybridoma cells.
  • Animal component-free
    Chemically defined
    GMP manufacture
With: 1176 mg/L Sodium bicarbonate
Without: Linoleic acid, L-Glutamine - add 5 ml/L (200 mM)