Order Entry
United States
Orders LinkContactUsLinkComponent
HEPES Buffer Solution 1 M

HEPES Buffer Solution 1 M

Liquid, 100 ml
Catalog # 77975-383
New Product
Supplier #:  9319-100ML
HEPES Buffer Solution 1 M
Catalog # 77975-383
New Product
CAS Number:  
HEPES Buffer Solution 1 M
HEPES Buffer Solution 1 M

Liquid, 100 ml

CAS Number:  
Price
$88.36
Log in for availability
Each(100ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Buffer to maintain pH between 7.2 and 7.6. Suitable for use in cell cultures outside of a CO₂ incubator

  • Animal-component free
  • Chemically defined
  • GMP manufacture

This formulation contains 238.3 g/L HEPES in water, pH 7.2 to 7.4 at room temperature.

Packaging: Store at 2 to 8 °C. Shelf life: 24 months from date of manufacture.

Specifications

Specifications

  • Size:
    100 ml
  • Format:
    Liquid
  • Cat. No.:
    77975-383
  • Calcium:
    No
  • Sodium bicarbonate:
    No
  • Phenol red:
    No

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.