Order Entry
United States
ContactUsLinkComponent
IS MAB-CD Chemically-Defined Medium
IS MAB-CD Chemically-Defined Medium
Catalog # 77975-319
New Product
CAS Number:  
undefined
IS MAB-CD Chemically-Defined Medium
Catalog # 77975-319
Supplier Number:  91104-1L
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Size:
    1 L
  • Calcium:
    No
  • Magnesium:
    No
  • HEPES:
    No
  • Sodium bicarbonate:
    No
  • Sodium pyruvate:
    No
  • Phenol red:
    No
  • Stable glutamine:
    No
  • L-Glutamine:
    No
  • Glucose level:
    Without
  • Media Base:
    Proprietary Mixture

Specifications

About this item

IS MAB-CD is a chemically-defined, animal component-free medium designed for the growth of hybridoma and myeloma cells lines for recombinant monoclonal antibody production.

  • Components are chemically-defined and non-animal in origin

    Contains no hydrolysates, extracts or other undefined components

    Lot-to-lot consistency

    Available in 1 L packaging

    Custom formulations, packaging, and powder configurations are available

    Without L-glutamine, add prior to use

FUJIFILM Irvine Scientific developed IS MAB-CD, a chemically-defined medium formulated specifically for the growth of hybridoma and myeloma cell lines for recombinant monoclonal antibody production. To address concerns relating to consistency, regulatory and downstream process issues, IS MAB-CD has been developed as a protein-free medium with components that are defined and of non-animal origin.

Packaging:

Store at 2 to 8 °C and protected from light

Shelf life of one year from date of manufacture