Specifications
- Media Format:Liquid
- Pack type:Bag
- Size:5 L basal media and 10 ml supplement
- Calcium:No
- Magnesium:No
- HEPES:No
- Sodium bicarbonate:No
- Sodium pyruvate:No
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:Without
- Media Base:Complete Medium
Specifications
About this item
The PRIME-XV MSC XSFM Dual component kits are xeno-free and serum-free media that enable a single-use, closed system format for MSC expansion in Quantum or other compatible hollow fiber bioreactors. The medium is formulated to deliver expansion and viability of MSCs over several passages in culture, while maintaining multipotency and functional characteristics.
Enables up to an 8-fold increase in MSC expansion (donor-dependent)
Yields 200×106 cells in 7 days of expansion with 90% and above viability
Supports expansion of MSCs; data derived from human adipose tissue
Maintains lot-to-lot consistency with xeno- and serum-free media
Simplifies and expedites process of MSC expansion for large scale solutions
Minimizes risk of contamination and cell stress, and increases efficiency
Does not contain antibiotics
Available with or without phenol red
PRIME-XV MSC XSFM Dual Component Kit with Phenol Red, 91149DC is a dual-component serum-free and xeno-free 5 L basal medium bag and a frozen supplement bag. When combined, the formulation is optimized for the maintenance and expansion of purified or enriched human mesenchymal stromal/stem cells (MSCs) in the Terumo Quantum or Quantum Flex Bioreactor, or other compatible bioreactors, under serum-free and xeno-free culture conditions. This product does not contain antibiotics. This medium is provided as a basal medium and supplement and should be mixed prior to use. It may also be used with additional cytokine/growth factors for desired application.