Order Entry
United States
ContactUsLinkComponent
Turbidity Wastewater QC Concentrate, Certified Reference Material
Turbidity Wastewater QC Concentrate, Certified Reference Material
Catalog # SXQCI-092
Supplier:  SPEX CERTIPREP LLC
New Product
CAS Number:  
undefined
Turbidity Wastewater QC Concentrate, Certified Reference Material
Catalog # SXQCI-092
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  QCI-092
New Product
CAS Number:  

Specifications

  • Concentration:
    2 to 30 NTU
  • Description:
    Turbidity wastewater QC concentrate, certified reference material, 21 ml
  • Shipping Conditions:
    Non-hazardous
  • Storage Temperature:
    Ambient
  • Physical Form:
    Liquid
  • Contents:
    Turbidity
  • Contents:
    Turbidity

Specifications

About this item

Meet or exceed performance criteria with QC samples.

  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation

    For US EPA Method 180.1 and SM 18/19th ED 2130 B

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016