Order Entry
United States
ContactUsLinkComponent
Anti-DNA Ligase IV/LIG4 Rabbit Monoclonal Antibody [clone: EPR16531]
Anti-DNA Ligase IV/LIG4 Rabbit Monoclonal Antibody [clone: EPR16531]
Supplier:  Abcam
New Product
Certificates
Anti-DNA Ligase IV/LIG4 Rabbit Monoclonal Antibody [clone: EPR16531]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB193353-1ML
  • ABCA_AB193353-100U
  • Antigen Symbol:
  • DNA Ligase IV/LIG4
  • DNA Ligase IV/LIG4
  • Antigen Name:
  • DNA ligase 4
  • DNA ligase 4
  • Antigen Synonyms:
  • DNA ligase IV|Polydeoxyribonucleotide synthase [ATP] 4|LIG4
  • DNA ligase IV|Polydeoxyribonucleotide synthase [ATP] 4|LIG4
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR16531
  • EPR16531
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 103971 Da
  • 103971 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Concentration:
  • 0.464 mg/ml
  • 0.464 mg/ml
  • Storage Temperature:
  • Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
  • Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • Shipped at +4 °C
  • Shipped at +4 °C
  • Tested Applications:
  • ICC/IF, IHC-P
  • ICC/IF, IHC-P

Specifications

Frequently Bought Together