Anti-S100A8 Mouse Monoclonal Antibody [clone: 7] (FITC)
Catalog # ABCA_AB275635-100T
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CAGA
- Antigen Symbol:S100A8
- Clonality:Monoclonal
- Clone:7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 Tests
- Form:Liquid
- Antigen Synonyms:CFAG|MRP8|Protein S100-A8|Calgranulin-A|Calprotectin L1L subunit|Cystic fibrosis antigen|Leukocyte L1 complex light chain|Migration inhibitory factor-related protein 8|S100 calcium-binding protein A8|Urinary stone protein band A|MRP-8|p8
- Molecular Weight:10835 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C. Do Not Freeze. Store In the Dark.
- Concentration:0.1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human S100A8.
- Tested Applications:Flow Cyt (Intra)
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
FITC Mouse monoclonal [07] to S100A8.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: S100A8
Clonality: Monoclonal
Clone: 7
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human