Anti-MS4A6A Rabbit Polyclonal Antibody
Catalog # ABCA_AB189983-50UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:4SPAN3
- Antigen Symbol:MS4A6A
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Antigen Synonyms:CD20L3|MS4A6|CDA01|MSTP090|Membrane-spanning 4-domains subfamily A member 6A|CD20 antigen-like 3|Four-span transmembrane protein 3
- Molecular Weight:26943 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human MS4A6A. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to MS4A6A - N-terminal.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MS4A6A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat; Human