Order Entry
United States
ContactUsLinkComponent
Anti-HCP1/PCFT Rabbit Polyclonal Antibody
Anti-HCP1/PCFT Rabbit Polyclonal Antibody
Catalog # ABCA_AB241949-100U
Supplier:  Abcam
New Product
Anti-HCP1/PCFT Rabbit Polyclonal Antibody
Catalog # ABCA_AB241949-100U
Supplier:  Abcam
Supplier Number:  AB241949-100UG
New Product

About this item

Rabbit polyclonal to HCP1/PCFT.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Astrin/Deepest
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human