Order Entry
United States
ContactUsLinkComponent
Anti-Laminin 2 alpha Rabbit Monoclonal Antibody [clone: EPR23924-60]
Anti-Laminin 2 alpha Rabbit Monoclonal Antibody [clone: EPR23924-60]
Supplier:  Abcam
New Product
Certificates
Anti-Laminin 2 alpha Rabbit Monoclonal Antibody [clone: EPR23924-60]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB275258-100U
  • ABCA_AB275258-1MG
  • Antigen Symbol:
  • Laminin 2 alpha
  • Laminin 2 alpha
  • Antigen Name:
  • Laminin subunit alpha-2
  • Laminin subunit alpha-2
  • Antigen Synonyms:
  • Laminin M chain|Laminin-12 subunit alpha|Laminin-2 subunit alpha|Laminin-4 subunit alpha|Merosin heavy chain|Lama2
  • Laminin M chain|Laminin-12 subunit alpha|Laminin-2 subunit alpha|Laminin-4 subunit alpha|Merosin heavy chain|Lama2
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR23924-60
  • EPR23924-60
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 343815 Da
  • 343815 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C
  • Store at 4 °C
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • IHC-Fr, IHC-P
  • IHC-Fr, IHC-P

Specifications

Frequently Bought Together