Anti-PSMA3 Rabbit Polyclonal Antibody
ABCA_AB264307-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:HC8
- Antigen Symbol:PSMA3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:PSC8|Proteasome subunit alpha type-3|Macropain subunit C8|Multicatalytic endopeptidase complex subunit C8|Proteasome component C8|Proteasome subunit alpha-7|alpha-7
- Molecular Weight:28433 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Concentration:0.2 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human PSMA3 aa 200 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to PSMA3.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PSMA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse