Anti-Histone H2A (acetyl K5) Rabbit Monoclonal Antibody [clone: EP856Y]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EP856Y] to Histone H2A (acetyl K5) - BSA and Azide free.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H2A
Clonality: Monoclonal
Clone: EP856Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCA_AB247302-1MG
- ABCA_AB247302-100U
- Antigen Symbol:
- Histone H2A
- Histone H2A
- Antigen Name:
- H2AFP
- H2AFP
- Antigen Synonyms:
- HIST1H2AG|H2AC13|H2AFC|HIST1H2AI|H2AC15|H2AFD|HIST1H2AK|H2AC16|H2AFI|HIST1H2AL|H2AC17|H2AFN|HIST1H2AM|H2AC11|Histone H2A type 1|H2A.1|Histone H2A/ptl|H2AK5ac
- HIST1H2AG|H2AC13|H2AFC|HIST1H2AI|H2AC15|H2AFD|HIST1H2AK|H2AC16|H2AFI|HIST1H2AL|H2AC17|H2AFN|HIST1H2AM|H2AC11|Histone H2A type 1|H2A.1|Histone H2A/ptl|H2AK5ac
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EP856Y
- EP856Y
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Modification:
- acetyl K5
- acetyl K5
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 14091 Da
- 14091 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C. Do not freeze.
- Store at +4 °C. Do not freeze.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Specifications
Frequently Bought Together