Anti-SLC17A1 Rabbit Monoclonal Antibody [clone: EPR9614]
: Abcam
New Product
Rabbit monoclonal [EPR9614] to SLC17A1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SLC17A1
Clonality: Monoclonal
Clone: EPR9614
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB151751-1ML
- ABCA_AB151751-100U
- Antigen Symbol:
- SLC17A1
- SLC17A1
- Antigen Name:
- NPT1
- NPT1
- Antigen Synonyms:
- Sodium-dependent phosphate transport protein 1|Na(+)/PI cotransporter 1|Na/Pi-4|Renal Na(+)-dependent phosphate cotransporter 1|Renal sodium-dependent phosphate transport protein 1|Sodium/phosphate cotransporter 1|Solute carrier family 17 member 1|Renal sodium-phosphate transport protein 1
- Sodium-dependent phosphate transport protein 1|Na(+)/PI cotransporter 1|Na/Pi-4|Renal Na(+)-dependent phosphate cotransporter 1|Renal sodium-dependent phosphate transport protein 1|Sodium/phosphate cotransporter 1|Solute carrier family 17 member 1|Renal sodium-phosphate transport protein 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR9614
- EPR9614
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 51132 Da
- 51132 Da
- Purification:
- Tissue culture supernatant
- Tissue culture supernatant
- Storage Temperature:
- Store at -20ºC
- Store at -20ºC
- Shipping Temperature:
- 4 °C
- 4 °C
Frequently Bought Together