Anti-ATG16L2 Rabbit Polyclonal Antibody
Catalog # ABCA_AB81596-100UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:WDR80
- Antigen Symbol:ATG16L2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Protein Atg16l2|APG16-like 2|Autophagy-related protein 16-2|WD repeat-containing protein 80
- Storage Buffer:Whole serum
- Molecular Weight:68998 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Store at −20 °C long term.
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human ATG16L2. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Whole antiserum
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to ATG16L2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ATG16L2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human