Anti-CRCP Rabbit Monoclonal Antibody [clone: EPR9670(B)]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR9670(B)] to CRCP.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CRCP
Clonality: Monoclonal
Clone: EPR9670(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCA_AB139264-1ML
- ABCA_AB139264-100U
- Antigen Symbol:
- CRCP
- CRCP
- Antigen Name:
- DNA-directed RNA polymerase III subunit RPC9
- DNA-directed RNA polymerase III subunit RPC9
- Antigen Synonyms:
- RNA polymerase III subunit C9|Calcitonin gene-related peptide-receptor component protein|CGRP-RCP|CGRP-receptor component protein|CGRPRCP|HsC17
- RNA polymerase III subunit C9|Calcitonin gene-related peptide-receptor component protein|CGRP-RCP|CGRP-receptor component protein|CGRPRCP|HsC17
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR9670(B)
- EPR9670(B)
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 16871 Da
- 16871 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at–20 ℃.
- Store at–20 ℃.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Specifications
Frequently Bought Together