Anti-RIP2 Rabbit Polyclonal Antibody
Catalog # ABCA_AB8428-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CARDIAK
- Antigen Symbol:RIP2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:RICK|UNQ277/PRO314/PRO34092|RIPK2|Receptor-interacting serine/threonine-protein kinase 2|CARD-containing interleukin-1 beta-converting enzyme-associated kinase|RIP-like-interacting CLARP kinase|Receptor-interacting protein 2|Tyrosine-protein kinase RIPK2|CARD-containing IL-1 beta ICE-kinase|RIP-2
- Storage Buffer:PBS
- Molecular Weight:61195 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Store at +4 °C.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human RIPK2 aa 1 - 50. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to RIP2.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RIP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human