Anti-c-Myc Rabbit Monoclonal Antibody [clone: Y69]
: Abcam
New Product
Rabbit monoclonal [Y69] to c-Myc - ChIP Grade.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: Y69
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- ABCA_AB32072-1ML
- ABCA_AB32072-100UL
- ABCA_AB32072-20UL
- Antigen Symbol:
- c-Myc
- c-Myc
- c-Myc
- Antigen Name:
- BHLHE39
- BHLHE39
- BHLHE39
- Antigen Synonyms:
- MYC|Myc proto-oncogene protein|Class E basic helix-loop-helix protein 39|Proto-oncogene c-Myc|Transcription factor p64
- MYC|Myc proto-oncogene protein|Class E basic helix-loop-helix protein 39|Proto-oncogene c-Myc|Transcription factor p64
- MYC|Myc proto-oncogene protein|Class E basic helix-loop-helix protein 39|Proto-oncogene c-Myc|Transcription factor p64
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- Y69
- Y69
- Y69
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 48804 Da
- 48804 Da
- 48804 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Storage Buffer:
- BSA, glycerol, sodium chloride, tris glycine
- BSA, glycerol, sodium chloride, tris glycine
- BSA, glycerol, sodium chloride, tris glycine
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested Applications:
- ChIC/CUT&RUN-seq, ChIP-seq, Flow Cyt (Intra), ICC/IF, IHC-P
- ChIC/CUT&RUN-seq, ChIP-seq, Flow Cyt (Intra), ICC/IF, IHC-P
- ChIC/CUT&RUN-seq, ChIP-seq, Flow Cyt (Intra), ICC/IF, IHC-P
Frequently Bought Together