Anti-NG2 Rabbit Monoclonal Antibody [clone: EPR23752-147] (Alexa Fluor® 647)
ABCA_AB307168-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:An2
- Antigen Symbol:NG2
- Clonality:Monoclonal
- Clone:EPR23752-147
- Conjugation:Alexa Fluor® 647
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Kiaa4232|Cspg4|Chondroitin sulfate proteoglycan 4|Chondroitin sulfate proteoglycan NG2|Proteoglycan
- Molecular Weight:252309 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Purification:Protein A Affinity purification
- Packaging:Vial
Alexa Fluor® 647 Rabbit monoclonal [EPR23752-147] to NG2.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: NG2
Clonality: Monoclonal
Clone: EPR23752-147
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse