Anti-AMPK alpha 1 Rabbit Monoclonal Antibody [clone: EPR26792-76]
: Abcam
New Product
Rabbit monoclonal [EPR26792-76] to AMPK alpha 1 - BSA and Azide free (Capture).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: AMPK alpha 1
Clonality: Monoclonal
Clone: EPR26792-76
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- ABCA_AB316833-100U
- ABCA_AB316833-1MG
- Antigen Symbol:
- AMPK alpha 1
- AMPK alpha 1
- Antigen Name:
- AMPK1
- AMPK1
- Antigen Synonyms:
- PRKAA1|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK subunit alpha-1|Acetyl-CoA carboxylase kinase|Hydroxymethylglutaryl-CoA reductase kinase|Tau-protein kinase PRKAA1|ACACA kinase|HMGCR kinase
- PRKAA1|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK subunit alpha-1|Acetyl-CoA carboxylase kinase|Hydroxymethylglutaryl-CoA reductase kinase|Tau-protein kinase PRKAA1|ACACA kinase|HMGCR kinase
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR26792-76
- EPR26792-76
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 64009 Da
- 64009 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- sELISA
- sELISA
Frequently Bought Together